• USA Home
  • Search Results

  • rhodium10291744016611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rhodium

42 Products
Image of Rhodium
CAS Number:
Empirical Formula (Hill Notation):
 Rh
Molecular Weight:
 102.91
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Composite Dimension
  Physical Form
  Manufacturer designation
Show More

Product #
Description
204218 Rhodium, powder, 99.95% trace metals basis,
357340 Rhodium, foil, thickness 0.025 mm, 99.9% trace metals basis,
GF37380089 Rhodium, wire reel, 0.2m, diameter 0.25mm, hard, 99.9%,
GF30467998 Rhodium, foil, thickness 0.025 mm, size 10 × 10 mm, purity 99.9%,
GF31954866 Rhodium, foil, thickness 0.025 mm, size 25 × 25 mm, purity 99.9%,
GF52123788 Rhodium, wire, 1.0 mm diameter, length 0.025 m, purity 99.9%,
GF44569907 Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%,
GF80841146 Rhodium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%,
GF40539405 Rhodium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%,
GF20569697 Rhodium, foil, 12mm disks, thickness 0.5mm, as rolled, 99.9%,
GF36379345 Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%,
GF57675561 Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%,
GF12568013 Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, x-ray quality, 99.9%,
GF96356559 Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%,
GF04978963 Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%,
GF17514312 Rhodium, foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%,
GF35442727 Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%,
GF49036204 Rhodium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.85%,
GF87742659 Rhodium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.85%,
GF15499413 Rhodium, foil, not light tested, 25x25mm, thickness 0.006mm, as rolled, 99.9%,
GF24911726 Rhodium, foil, not light tested, 25x25mm, thickness 0.012mm, as rolled, 99.9%,
GF82366342 Rhodium, foil, not light tested, 50x50mm, thickness 0.00025mm, permanent polyester support, 99.9%,
GF22634120 Rhodium, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.85%,
GF33789550 Rhodium, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.85%,
GF81993910 Rhodium, foil, not light tested, 50x50mm, thickness 0.006mm, as rolled, 99.9%,
GF71687357 Rhodium, microfoil, disks, 10mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF94930667 Rhodium, microfoil, disks, 10mm, thinness 0.25μm, specific density 310μg/cm2, permanent mylar 3.5μm support, 99.99%,
GF49946417 Rhodium, microfoil, disks, 10mm, thinness 0.50μm, specific density 620μg/cm2, permanent mylar 3.5μm support, 99.99%,
GF14938005 Rhodium, microfoil, disks, 25mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF91583264 Rhodium, microfoil, disks, 25mm, thinness 0.25μm, specific density 310μg/cm2, permanent mylar 3.5μm support, 99.99%,
GF64213680 Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%,
GF45819995 Rhodium, wire reel, 0.05m, diameter 0.25mm, hard, 99.9%,
GF74452316 Rhodium, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.9%,
GF83497652 Rhodium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%,
GF26710680 Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%,
GF27499045 Rhodium, wire reel, 0.1m, diameter 0.25mm, hard, 99.9%,
GF06539979 Rhodium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%,
GF37207166 Rhodium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9%,
GF38835737 Rhodium, wire reel, 0.2m, diameter 0.12mm, as drawn, 99.9%,
GF17496973 Rhodium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%,
GF54466937 Rhodium, wire reel, 0.5m, diameter 0.12mm, as drawn, 99.9%,
GF72510364 Rhodium, wire reel, 0.5m, diameter 0.25mm, hard, 99.9%,