• USA Home
  • Search Results

  • rhodiumiiichloride209261004907711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rhodium(III) chloride

1 Products
Image of Rhodium(III) chloride
CAS Number:
Linear Formula:
 RhCl3
Molecular Weight:
 209.26
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More