• USA Home
  • Search Results

  • rhodiumiitrifluoroacetatedimer657873112695111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rhodium(II) trifluoroacetate dimer

1 Products
Synonym:   Trifluoroacetic acid rhodium(II) salt dimer
Image of Rhodium(II) trifluoroacetate dimer
CAS Number:
Linear Formula:
 [(CF3COO)2Rh]2
Molecular Weight:
 657.87
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More