• USA Home
  • Search Results

  • ribonucleaseafrombovinepancreas12345900199411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ribonuclease A from bovine pancreas

8 Products
Synonym:   Pancreatic Ribonuclease, RNase A, Ribonucleate 3′-pyrimidinooligonucleotidohydrolase
Image of Ribonuclease A from bovine pancreas
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.1.27.5  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.53
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  Color
  Ice Shipping
  Type
  Molecular weight
  Assay
Show More

Product #
Description
R6513 Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized,
R4642 Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0),
R4875 Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein,
R5503 Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein,
R5125 Ribonuclease A from bovine pancreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein,
R5000 Ribonuclease A from bovine pancreas, Type II-A, ≥60% (SDS-PAGE), 50-150 Kunitz units/mg solid,
R5500 Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein,
R5250 Ribonuclease A from bovine pancreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein,