• USA Home
  • Search Results

  • ricinoleicacid2984614122011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ricinoleic acid

2 Products
Synonym:   (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid, Ricinelaidic acid
Image of Ricinoleic acid
CAS Number:
Linear Formula:
 CH3(CH2)5CH(OH)CH2CH=CH(CH2)7COOH
Molecular Weight:
 298.46
 Beilstein/REAXYS Number:
 1727813
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
  Physical Form
  Special Grade
  Application(s)
  Ice Shipping
  Assay
Show More