• USA Home
  • Search Results

  • rifampicin822941329246111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rifampicin

4 Products
Synonym:   3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampin, Rifamycin AMP
Image of Rifampicin
CAS Number:
Empirical Formula (Hill Notation):
 C43H58N4O12
Molecular Weight:
 822.94
 Beilstein Registry Number:
 5723476
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Anal. Standard Form
  Product Line
  pK-Values
  Titertest
  Color
  Assay
  Mode of Action
  Physical Form
  Solubility
Show More