• USA Home
  • Search Results

  • rifaximin785888062181411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rifaximin

3 Products
Synonym:   4-Deoxy-4′-methylpyrido[1′,2′-1,2]imidazo[5,4-c]rifamycin SV, Rifacol
Image of Rifaximin
CAS Number:
Empirical Formula (Hill Notation):
 C43H51N3O11
Molecular Weight:
 785.88
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Activity Spectrum
  Titertest
  Color
Show More