• USA Home
  • Search Results

  • rmandelicacid1521561171211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(−)-Mandelic acid

3 Products
Synonym:   (R)-α-Hydroxyphenylacetic acid
Image of (<I>R</I>)-(−)-Mandelic acid
CAS Number:
Linear Formula:
 C6H5CH(OH)CO2H
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 2691094
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More