• USA Home
  • Search Results

  • robinin7406630119911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Robinin

2 Products
Synonym:   Kaempferol 3-O-robinoside-7-O-rhamnoside
Image of Robinin
CAS Number:
Empirical Formula (Hill Notation):
 C33H40O19
Molecular Weight:
 740.66
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More