• USA Home
  • Search Results

  • rocuroniumbromide6096811930291911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rocuronium bromide

3 Products
Image of Rocuronium bromide
CAS Number:
Empirical Formula (Hill Notation):
 C32H53BrN2O4
Molecular Weight:
 609.68
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Manufacturer designation
Show More