• USA Home
  • Search Results

  • rosavin428438495492711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rosavin

3 Products
Synonym:   (2E)-3-Phenylprop-2-en-1-yl 6-O-α-L-arabinopyranosyl-β-D-glucopyranoside, (2R,3R,4S,5S,6R)-2-[(E)-3-Phenylprop-2-enoxy]-6-([(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxymethyl)oxane-3,4,5-triol, (E)-3-Phenyl-2-propenyl 6-O-a-L-arabinopyranosyl-β-D-glucopyranoside, Rosavidin
Image of Rosavin
CAS Number:
Empirical Formula (Hill Notation):
 C20H28O10
Molecular Weight:
 428.43
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More