• USA Home
  • Search Results

  • roscovitine3544518669246611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Roscovitine

1 Products
Synonym:   2-(R)-[[9-(1-Methylethyl)-6-[(phenylmethyl)amino]-9H-purin-2-yl]amino]-1-butanol, 6-(Benzylamino)-2(R)-[[1-(hydroxymethyl)propyl]amino]-9-isopropylpurine
Image of Roscovitine
CAS Number:
Empirical Formula (Hill Notation):
 C19H26N6O
Molecular Weight:
 354.45
 MDL number:
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
R7772 Roscovitine, ≥98% (TLC),