• USA Home
  • Search Results

  • rotigotinehydrochloride3519312557293211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rotigotine hydrochloride

2 Products
Synonym:   (−)-(S)-5,6,7,8-Tetrahydro-6-(propyl(2-(2-thienyl)ethyl)amino)-1-naphthol hydrochloride, (−)-N 0437, N 0923, Neupro, SPM 962
Image of Rotigotine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H25NOS·HCl
Molecular Weight:
 351.93
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More