• USA Home
  • Search Results

  • rphenylephrinehydrochloride203676176711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(−)-Phenylephrine hydrochloride

2 Products
Synonym:   (R)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride
Image of (<I>R</I>)-(−)-Phenylephrine hydrochloride
CAS Number:
Linear Formula:
 HOC6H4CH(CH2NHCH3)OH·HCl
Molecular Weight:
 203.67
 Beilstein/REAXYS Number:
 4158948
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More