• USA Home
  • Search Results

  • rtolbinap678789964628311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-Tol-BINAP

1 Products
Synonym:   (R)-(+)-2,2′-Bis(di-p-tolylphosphino)-1,1′-binaphthyl
Image of (<I>R</I>)-Tol-BINAP
CAS Number:
Empirical Formula (Hill Notation):
 C48H40P2
Molecular Weight:
 678.78
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
Show More

Product #
Description
693049 (R)-Tol-BINAP,