• USA Home
  • Search Results

  • rubpm3cl2646466503488011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

[Ru(bpm)3][Cl]2

1 Products
Synonym:   Dichlorotris(2,2′-bipyrimidine-N1,N1′)ruthenium(II), Tris(2,2′-bipyrimide)ruthenium(II) dichloride
Image of [Ru(bpm)<SUB>3</SUB>][Cl]<SUB>2</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C24H18Cl2N12Ru
Molecular Weight:
 646.46
 NACRES:
 NA.22
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
747785 [Ru(bpm)3][Cl]2,