• USA Home
  • Search Results

  • rutheniumiiichloride207431004908811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ruthenium(III) chloride

1 Products
Image of Ruthenium(III) chloride
CAS Number:
Linear Formula:
 RuCl3
Molecular Weight:
 207.43
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More