• USA Home
  • Search Results

  • rutheniumiiichloridehydrate20743anhydrousbasis1489867011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ruthenium(III) chloride hydrate

4 Products
Synonym:   Ruthenium trichloride
Image of Ruthenium(III) chloride hydrate
CAS Number:
Linear Formula:
 RuCl3 · xH2O
Molecular Weight:
 207.43 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More