• USA Home
  • Search Results

  • rutheniumivoxidehydrate13307anhydrousbasis3274079711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ruthenium(IV) oxide hydrate

2 Products
Synonym:   Hydrous ruthenium oxide, Ruthenium dioxide hydrate
Image of Ruthenium(IV) oxide hydrate
CAS Number:
Linear Formula:
 RuO2 · xH2O
Molecular Weight:
 133.07 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More