• USA Home
  • Search Results

  • rutheniumred786351110372311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ruthenium Red

3 Products
Synonym:   Ammoniated ruthenium oxychloride
Image of Ruthenium Red
CAS Number:
Linear Formula:
 [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6
Molecular Weight:
 786.35
 Colour Index Number:
 77800
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Absorption
  Physical Form
  Solubility
  Assay
Show More