• USA Home
  • Search Results

  • rutintrihydrate6645625024975311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rutin trihydrate

3 Products
Synonym:   Quercetin-3-rutinoside trihydrate, Vitamin P trihydrate
Image of Rutin trihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C27H30O16 · 3H2O
Molecular Weight:
 664.56
 Beilstein/REAXYS Number:
 75455
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Manufacturer designation
  Assay
  Special Grade
Show More