• USA Home
  • Search Results

  • s22bisdiphenylphosphino11binaphthalene622677618956511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene

1 Products
Synonym:   (S)-(−)-(1,1′-Binaphthalene-2,2′-diyl)bis(diphenylphosphine), (S)-(−)-BINAP
Image of (<I>S</I>)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene
CAS Number:
Linear Formula:
 [(C6H5)2PC10H6-]2
Molecular Weight:
 622.67
 Beilstein/REAXYS Number:
 5321443
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More