• USA Home
  • Search Results

  • s22dimethoxy11binaphthalene314387564087811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(−)-2,2′-Dimethoxy-1,1′-binaphthalene

1 Products
Synonym:   (S)-(−)-2,2′-Dimethoxy-1,1′-binaphthyl, (S)-1,1′-Bi-2-naphthyl dimethyl ether
Image of (<I>S</I>)-(−)-2,2′-Dimethoxy-1,1′-binaphthalene
CAS Number:
Empirical Formula (Hill Notation):
 C22H18O2
Molecular Weight:
 314.38
 Beilstein/REAXYS Number:
 4704232
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More