• USA Home
  • Search Results

  • s2methylcbsoxazaborolidinesolution2771711202281811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(−)-2-Methyl-CBS-oxazaborolidine solution

2 Products
Synonym:   α,α-Diphenyl-L-prolinol methylboronic acid cycl-amide ester, (S)-1-Methyl-3,3-diphenyl-tetrahydro-pyrrolo[1,2c][1,3,2]oxazaborole, (S)-3,3-Diphenyl-1-methyltetrahydro-3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole
Image of (<I>S</I>)-(−)-2-Methyl-CBS-oxazaborolidine solution
CAS Number:
Empirical Formula (Hill Notation):
 C18H20BNO
Molecular Weight:
 277.17
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More