• USA Home
  • Search Results

  • s4amino2hydroxybutyricacid119124037151511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(−)-4-Amino-2-hydroxybutyric acid

1 Products
Image of (<I>S</I>)-(−)-4-Amino-2-hydroxybutyric acid
CAS Number:
Linear Formula:
 H2NCH2CH2CH(OH)CO2H
Molecular Weight:
 119.12
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More