• USA Home
  • Search Results

  • s4benzyl2oxazolidinone177209071932711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-4-Benzyl-2-oxazolidinone

1 Products
Image of (<I>S</I>)<WBR>-<WBR>4-<WBR>Benzyl-<WBR>2-<WBR>oxazolidinone
CAS Number:
Empirical Formula (Hill Notation):
 C10H11NO2
Molecular Weight:
 177.20
 Beilstein/REAXYS Number:
 3649667
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More