• USA Home
  • Search Results

  • s4isopropyl2oxazolidinethione1452210449908311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(−)-4-Isopropyl-2-oxazolidinethione

1 Products
Image of (<I>S</I>)-(−)-4-Isopropyl-2-oxazolidinethione
CAS Number:
Empirical Formula (Hill Notation):
 C6H11NOS
Molecular Weight:
 145.22
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More