• USA Home
  • Search Results

  • sD1aminoethylphosphonicacid125066606876611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(+)-1-Aminoethylphosphonic acid

1 Products
Synonym:   D-(+)-1-Aminoethylphosphonic acid, D-Ala(P)
Image of (<I>S</I>)<WBR>-<WBR>(+)<WBR>-<WBR>1-<WBR>Aminoethylphosphonic acid
CAS Number:
Linear Formula:
 CH3CH(NH2)P(O)(OH)2
Molecular Weight:
 125.06
 Beilstein/REAXYS Number:
 4291033
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More