• USA Home
  • Search Results

  • sD1indanol134182550132011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(+)-1-Indanol

1 Products
Synonym:   (S)-(+)-1-Hydroxyindan
Image of (<I>S</I>)-(+)-1-Indanol
CAS Number:
Empirical Formula (Hill Notation):
 C9H10O
Molecular Weight:
 134.18
 Beilstein/REAXYS Number:
 2206709
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
323128 (S)-(+)-1-Indanol, 99%,