• USA Home
  • Search Results

  • sD2chloro1phenylethanol156617011105611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(+)-2-Chloro-1-phenylethanol

1 Products
Synonym:   (S)-(+)-α-(Chloromethyl)benzenemethanol
Image of (<I>S</I>)<WBR>-<WBR>(+)<WBR>-<WBR>2-<WBR>Chloro-<WBR>1-<WBR>phenylethanol
CAS Number:
Linear Formula:
 C6H5CH(CH2Cl)OH
Molecular Weight:
 156.61
 Beilstein/REAXYS Number:
 3648988
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More