• USA Home
  • Search Results

  • sDamethoxyphenylaceticacid166172616426111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(+)-α-Methoxyphenylacetic acid

1 Products
Synonym:   O-Methyl-L-mandelic acid
Image of (<I>S</I>)-(+)-α-Methoxyphenylacetic acid
CAS Number:
Linear Formula:
 C6H5CH(OCH3)CO2H
Molecular Weight:
 166.17
 Beilstein/REAXYS Number:
 2361718
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More