• USA Home
  • Search Results

  • sDcamptothecin34835768903411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(+)-Camptothecin

1 Products
Image of (<I>S</I>)-(+)-Camptothecin
CAS Number:
Empirical Formula (Hill Notation):
 C20H16N2O4
Molecular Weight:
 348.35
 Beilstein/REAXYS Number:
 631069
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Mode of Action
Show More