• USA Home
  • Search Results

  • sabouraud4glucoseagar1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sabouraud 4% Glucose Agar

1 Products
 NACRES:
 NA.74
Popular Documents:  Datasheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Media Suitability
  Shelf Life
  Special Grade
  pH Value
  Chemical Composition
Show More