• USA Home
  • Search Results

  • saccharin183188107211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Saccharin

7 Products
Synonym:   2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide, 2-Sulfobenzoic acid imide, o-Benzoic sulfimide
Image of Saccharin
CAS Number:
Empirical Formula (Hill Notation):
 C7H5NO3S
Molecular Weight:
 183.18
 Beilstein/REAXYS Number:
 6888
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Agency/Method
  Application(s)
  Manufacturer designation
  Melting Point
  Anal. Standard Form
  Physical Form
  Assay
  Special Grade
Show More