• USA Home
  • Search Results

  • sakuranetin2862852029611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sakuranetin

2 Products
Synonym:   4′-5-Dihydroxy-7-methoxyflavanone, 7-O-Methylnaringenin, Naringenin 7-O-methyl ether
Image of Sakuranetin
CAS Number:
Empirical Formula (Hill Notation):
 C16H14O5
Molecular Weight:
 286.28
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More