• USA Home
  • Search Results

  • salbutamol239311855994911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Salbutamol

4 Products
Synonym:   α-[(tert-Butylamino)methyl]-4-hydroxy-m-xylene-α,α′-diol, Albuterol
Image of Salbutamol
CAS Number:
Empirical Formula (Hill Notation):
 C13H21NO3
Molecular Weight:
 239.31
 Beilstein/REAXYS Number:
 6405698
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More