• USA Home
  • Search Results

  • salbutamolhemisulfatesalt288355102270911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Salbutamol hemisulfate salt

2 Products
Synonym:   α-([t-Butylamino]methyl)-4-hydroxy-m-xylene-α,α′-diol, Albuterol
Image of Salbutamol hemisulfate salt
CAS Number:
Linear Formula:
 C13H21NO3 · 1/2H2SO4
Molecular Weight:
 288.35
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More