• USA Home
  • Search Results

  • salicylicacid138126972711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Salicylic acid

18 Products
Synonym:   2-Hydroxybenzoic acid, Salicylic acid
Image of Salicylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C7H6O3
Molecular Weight:
 138.12
 EC Index Number
 200-712-3
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Suitability
  Purity Grade
  Product Line
  Loss
  pH Value
  Vapor pressure
  Transition temperature
  Titertest
  Density
  Bulk Density
  Autoignition temp
  Physical Form
  Anal. Standard Form
  Special Grade
  Solubility
  Potency
  Melting Point
  Assay
Show More

Product #
Description
S5922 Salicylic acid, BioXtra, ≥99.0%,
84210 Salicylic acid, puriss. p.a., ≥99.0% (T),
W398500 Salicylic acid, ≥99%, FG,
S7401 Salicylic acid, suitable for plant cell culture,
27301 Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance),
105910 Salicylic acid, ReagentPlus®, ≥99%,
84211 Salicylic acid, tested according to Ph Eur,
52341 Salicylic acid, certified reference material, TraceCERT®,
1.00635 Salicylic acid, fine powder EMPROVE® ESSENTIAL Ph Eur,BP,USP,
1.00631 Salicylic acid, EMPROVE® ESSENTIAL Ph Eur,BP,USP,
8.18731 Salicylic acid, for synthesis,
PHL80529 Salicylic acid, phyproof® Reference Substance,
1609002 Salicylic acid, United States Pharmacopeia (USP) Reference Standard,
PHR1013 Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material,
BP775 Salicylic acid, British Pharmacopoeia (BP) Reference Standard,
S0200000 Salicylic acid, European Pharmacopoeia (EP) Reference Standard,
S-019 Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®,
247588 Salicylic acid, ACS reagent, ≥99.0%,