• USA Home
  • Search Results

  • samarium15036744019911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Samarium

21 Products
Synonym:   Samarium
Image of Samarium
CAS Number:
Empirical Formula (Hill Notation):
 Sm
Molecular Weight:
 150.36
 EC Index Number
 231-128-7
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Weight
  Particle Dimension
  Composite Dimension
  Manufacturer designation
  Physical Form
Show More

Product #
Description
261203 Samarium, −40 mesh, 99% trace rare earth metals basis,
693731 Samarium, foil, thickness 1 mm, 99.9% trace rare earth metals basis,
263184 Samarium, ingot, 99.9% trace rare earth metals basis,
8.14479 Samarium, powder for synthesis,
GF93054865 Samarium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%,
GF98883848 Samarium, rod, 100mm, diameter 6.35mm, cast, 99%,
GF65804238 Samarium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%,
GF25529407 Samarium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%,
GF55991076 Samarium, rod, 50mm, diameter 6.35mm, cast, 99%,
GF52166943 Samarium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%,
GF21658642 Samarium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%,
GF75301668 Samarium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%,
GF31858454 Samarium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%,
GF24936891 Samarium, foil, thickness 0.7 mm, 15 mm diameter, purity 99%,
GF39720641 Samarium, powder, max. particle size 500 micron, weight 50 g, purity 99.9%,
GF40930770 Samarium, lump, 25 mm max. lump size, weight 10 g, purity 99.9%,
GF47928460 Samarium, foil, thickness 0.5 mm, 15 mm diameter, purity 99%,
GF57343204 Samarium, lump, 25 mm max. lump size, weight 50 g, purity 99.9%,
GF74504707 Samarium, lump, 25 mm max. lump size, weight 20 g, purity 99.9%,
GF82306548 Samarium, powder, 500 max. part. size (micron), weight 10 g, purity 99.9%,
GF88317760 Samarium, powder, 500 max. part. size (micron), weight 20 g, purity 99.9%,