• USA Home
  • Search Results

  • samethoxyatrifluoromethylphenylaceticacid234171725771511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

2 Products
Synonym:   (−)-MTPA, Mosher’s acid
Image of (<I>S</I>)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
CAS Number:
Linear Formula:
 C6H5C(OCH3)(CF3)CO2H
Molecular Weight:
 234.17
 Beilstein/REAXYS Number:
 4684048
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More