• USA Home
  • Search Results

  • sandwhitequartz60081480860711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sand, white quartz

2 Products
Synonym:   Quartz, Silicon dioxide
Image of Sand, white quartz
CAS Number:
Linear Formula:
 SiO2
Molecular Weight:
 60.08
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Restriction
  Special Grade
Show More