• USA Home
  • Search Results

  • sarafloxacinhydrochloridehydrate42183anhydrousbasis91296876anhydrous11

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sarafloxacin hydrochloride hydrate

1 Products
Image of Sarafloxacin hydrochloride hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C20H17F2N3O3 · HCl · xH2O
Molecular Weight:
 421.83 (anhydrous basis)
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More