• USA Home
  • Search Results

  • sarcosine890910797111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sarcosine

4 Products
Synonym:   Methylaminoacetic acid, N-Methylglycine, Methylaminoacetic acid, N-Methylglycine
Image of Sarcosine
CAS Number:
Empirical Formula (Hill Notation):
 C3H7NO2
Molecular Weight:
 89.09
 EC Index Number
 203-538-6
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More