• USA Home
  • Search Results

  • sbcitronellol15627754051411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-(−)-β-Citronellol

3 Products
Synonym:   β-Rhodinol, (S)-3,7-Dimethyl-6-octen-1-ol
Image of (<I>S</I>)-(−)-β-Citronellol
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
Molecular Weight:
 156.27
 Beilstein Registry Number:
 1721505
 EC Number:
 FEMA Number:
 2309
 Flavis number:
 2.229
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More