• USA Home
  • Search Results

  • sbmethylphenethylamine135211759679111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-β-Methylphenethylamine

1 Products
Synonym:   (S)-2-Phenyl-1-propylamine
Image of (<I>S</I>)-β-Methylphenethylamine
CAS Number:
Linear Formula:
 C6H5CH(CH3)CH2NH2
Molecular Weight:
 135.21
 Beilstein Registry Number:
 3195645
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More