• USA Home
  • Search Results

  • schaftoside564495193832011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Schaftoside

2 Products
Synonym:   8-α-L-Arabinopyranosyl-6-β-D-glucopyranosyl-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Apigenin 8-C-α-L-arabinoside 6-C-β-D-glucoside, Shaftoside
Image of Schaftoside
CAS Number:
Empirical Formula (Hill Notation):
 C26H28O14
Molecular Weight:
 564.49
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More