• USA Home
  • Search Results

  • sciadopitysin5805452134611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sciadopitysin

1 Products
Synonym:   4′,4′′′,7-Trimethylamentoflavone, 5,7-Dihydroxy-8-(5-(5-hydroxy-7-methoxy-4-oxo-4H-1-benzopyran-2-yl)-2-methoxyphenyl)-2-(4-methoxyphenyl)-4-benzopyrone, Amentoflavone-7,4′,4”′- trimethylether
Image of Sciadopitysin
CAS Number:
Empirical Formula (Hill Notation):
 C33H24O10
Molecular Weight:
 580.54
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More