• USA Home
  • Search Results

  • scisverbenol152231888104411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(S)-cis-Verbenol

1 Products
Synonym:   (1S,2S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-ol
Image of (<I>S</I>)-<I>cis</I>-Verbenol
CAS Number:
Empirical Formula (Hill Notation):
 C10H16O
Molecular Weight:
 152.23
 Beilstein/REAXYS Number:
 2206582
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
247065 (S)-cis-Verbenol, 95%,