• USA Home
  • Search Results

  • sclareol3085051503711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sclareol

4 Products
Synonym:   (1R,2R,8aS)-Decahydro-1-(3-hydroxy-3-methyl-4-pentenyl)-2,5,5,8a-tetramethyl-2-naphthol
Image of Sclareol
CAS Number:
Empirical Formula (Hill Notation):
 C20H36O2
Molecular Weight:
 308.50
 Beilstein/REAXYS Number:
 2054148
 EC Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Shelf Life
  Special Grade
  Titertest
  Assay
Show More