• USA Home
  • Search Results

  • scopolaminehydrobromide43831653368211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Scopolamine hydrobromide

3 Products
Synonym:   (−)-Scopolamine hydrobromide trihydrate, Hyoscine hydrobromide, Scopine tropate
Image of Scopolamine hydrobromide
CAS Number:
Empirical Formula (Hill Notation):
 C17H21NO4 · HBr · 3H2O
Molecular Weight:
 438.31
 Beilstein Registry Number:
 6100669
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More